Assay Details
| Gene Symbol: Ahctf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026375.2 | NP_080651.2 | 22 - 23 | 3475 | 82 | |
| GenBank mRNA | AB059278.1 | - | 22 - 23 | 3131 | 82 | |
| BC138343.1 | - | 22 - 23 | 3118 | 8861363 | 82 | |
| BC138344.1 | - | 22 - 23 | 3118 | 8861364 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ahctf1
Genomic Map