Assay Details
| Gene Symbol: Gm6402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030688.1 | 1 | 154 | 59 | ||
| GenBank mRNA | CN670942.1 | - | 1 | 59 | ||
| Gene Symbol: Rps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009092.3 | NP_033118.1 | 1 - 2 | 37 | 59 | |
| GenBank mRNA | AK007992.1 | - | 1 - 2 | 45 | 59 | |
| AK010647.1 | - | 1 - 2 | 39 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007992.1, AK010647.1, NM_009092.3 transcripts.
Set Membership: |
|

Gm6402
Genomic Map