Assay Details
| Gene Symbol: Dlg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011807.3 | NP_035937.2 | 4 - 5 | 524 | 117 | |
| GenBank mRNA | AF388675.1 | - | 4 - 5 | 303 | 117 | |
| AK046525.1 | - | 3 - 4 | 564 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlg2
Genomic Map