Assay Details
| Gene Symbol: Ticrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029835.1 | NP_084111.1 | 18 - 19 | 3291 | 120 | |
| GenBank mRNA | AK051639.1 | - | 18 - 19 | 3254 | 120 | |
| AK054492.1 | - | 18 - 19 | 3258 | 120 | ||
| AK137597.1 | - | 18 - 19 | 3296 | 120 | ||
| BC158103.1 | - | 18 - 19 | 3255 | 9088180 | 120 | |
More Information
Set Membership: |
|

Ticrr
Genomic Map