Assay Details
| Gene Symbol: Ticrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029835.1 | NP_084111.1 | 8 - 9 | 2197 | 155 | |
| GenBank mRNA | AK041726.1 | - | 8 - 9 | 2147 | 155 | |
| AK051639.1 | - | 8 - 9 | 2160 | 155 | ||
| AK054492.1 | - | 8 - 9 | 2147 | 155 | ||
| AK137597.1 | - | 8 - 9 | 2163 | 155 | ||
| AK157802.1 | - | 7 - 8 | 3754 | 155 | ||
| BC158103.1 | - | 8 - 9 | 2161 | 9088180 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ticrr
Genomic Map