Assay Details
| Gene Symbol: Ticrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029835.1 | NP_084111.1 | 14 - 15 | 2856 | 69 | |
| GenBank mRNA | AK051639.1 | - | 14 - 15 | 2819 | 69 | |
| AK054492.1 | - | 14 - 15 | 2806 | 69 | ||
| AK137597.1 | - | 14 - 15 | 2861 | 69 | ||
| BC158103.1 | - | 14 - 15 | 2820 | 9088180 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ticrr
Genomic Map