Assay Details
| Gene Symbol: Sv2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109753.1 | NP_001103223.1 | 3 - 4 | 1194 | 100 | |
| NM_153579.4 | NP_705807.2 | 3 - 4 | 1401 | 100 | ||
| GenBank mRNA | AK029736.1 | - | 4 - 5 | 1226 | 100 | |
| AK043830.1 | - | 3 - 4 | 1121 | 100 | ||
| AK122359.1 | - | 2 - 3 | 346 | 100 | ||
| AK139478.1 | - | 3 - 4 | 1096 | 100 | ||
| AK140104.1 | - | 3 - 4 | 1114 | 100 | ||
| AK147261.1 | - | 3 - 4 | 1188 | 100 | ||
| AK159028.1 | - | 3 - 4 | 1099 | 100 | ||
| BC060224.1 | - | 2 - 3 | 785 | 30356990 | 100 | |
| BC079568.1 | - | 3 - 4 | 1401 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sv2b
Genomic Map