Assay Details
| Gene Symbol: Sv2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109753.1 | NP_001103223.1 | 8 - 9 | 1779 | 132 | |
| NM_153579.4 | NP_705807.2 | 8 - 9 | 1986 | 132 | ||
| GenBank mRNA | AK029736.1 | - | 9 - 10 | 1811 | 132 | |
| AK043830.1 | - | 8 - 9 | 1706 | 132 | ||
| AK122359.1 | - | 7 - 8 | 931 | 132 | ||
| AK139478.1 | - | 8 - 9 | 1681 | 132 | ||
| AK140104.1 | - | 8 - 9 | 1699 | 132 | ||
| AK147261.1 | - | 8 - 9 | 1773 | 132 | ||
| AK159028.1 | - | 8 - 9 | 1684 | 132 | ||
| BC060224.1 | - | 7 - 8 | 1370 | 30356990 | 132 | |
| BC079568.1 | - | 8 - 9 | 1986 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sv2b
Genomic Map