Assay Details
| Gene Symbol: Mtmr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144843.4 | NP_659092.1 | 1 - 2 | 157 | 71 | |
| GenBank mRNA | AK076218.1 | - | 1 - 2 | 134 | 71 | |
| AK154859.1 | - | 1 - 2 | 77 | 71 | ||
| BC020019.1 | - | 1 - 2 | 91 | 3596732 | 71 | |
| BY274243.1 | - | 1 - 2 | 157 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076218.1, AK154859.1, BC020019.1, NM_144843.4 transcripts.
Set Membership: |
|

Mtmr6
Genomic Map