Assay Details
| Gene Symbol: Mtmr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144843.4 | NP_659092.1 | 4 - 5 | 595 | 79 | |
| GenBank mRNA | AB517623.1 | - | 4 - 5 | 462 | 79 | |
| AK076218.1 | - | 4 - 5 | 572 | 79 | ||
| AK154859.1 | - | 4 - 5 | 515 | 79 | ||
| BC020019.1 | - | 4 - 5 | 529 | 3596732 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr6
Genomic Map