Assay Details
| Gene Symbol: Cib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291276.1 | NP_001278205.1 | 1 - 2 | 297 | 68 | |
| GenBank mRNA | AK088450.1 | - | 1 - 2 | 112 | 68 | |
| BB843350.1 | - | 1 - 2 | 76 | 68 | ||
More Information
Set Membership: |
|

Cib1
Genomic Map