Assay Details
| Gene Symbol: A930005H10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027894.1 | 1 - 2 | 286 | 148 | ||
| GenBank mRNA | BF019418.1 | - | 1 - 2 | 286 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930005H10Rik| Gene Symbol: A930005H10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027894.1 | 1 - 2 | 286 | 148 | ||
| GenBank mRNA | BF019418.1 | - | 1 - 2 | 286 | 148 | |
Set Membership: |
|