Assay Details
| Gene Symbol: Rab30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029494.2 | NP_083770.2 | 3 - 4 | 318 | 109 | |
| GenBank mRNA | AB232626.1 | - | 2 - 3 | 181 | 109 | |
| AF399756.1 | - | 2 - 3 | 181 | 109 | ||
| AK142206.1 | - | 3 - 4 | 325 | 109 | ||
| BC017550.1 | - | 3 - 4 | 318 | 4503909 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab30
Genomic Map