Assay Details
| Gene Symbol: Adam28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048175.2 | NP_001041640.1 | 17 - 18 | 2068 | 60 | |
| NM_010082.2 | NP_034212.1 | 17 - 18 | 2068 | 60 | ||
| NM_183366.3 | NP_899222.1 | 17 - 18 | 2068 | 60 | ||
| GenBank mRNA | AF153350.1 | - | 17 - 18 | 1975 | 60 | |
| AF163290.1 | - | 17 - 18 | 2068 | 60 | ||
| AF163291.1 | - | 17 - 18 | 2068 | 60 | ||
| AF163292.1 | - | 17 - 18 | 2068 | 60 | ||
| AK136715.1 | - | 8 - 9 | 972 | 60 | ||
| BC058782.1 | - | 17 - 18 | 1943 | 6310763 | 60 | |
More Information
Set Membership: |
|

Adam28
Genomic Map