Assay Details
| Gene Symbol: Mmp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008608.3 | NP_032634.3 | 3 - 4 | 634 | 71 | |
| GenBank mRNA | AK040968.1 | - | 606 | 71 | ||
| AK088476.1 | - | 3 - 4 | 634 | 71 | ||
| AK134340.1 | - | 3 - 4 | 565 | 71 | ||
| AK138611.1 | - | 3 - 4 | 575 | 71 | ||
| AK149636.1 | - | 3 - 4 | 563 | 71 | ||
| AK149907.1 | - | 3 - 4 | 614 | 71 | ||
| AK149988.1 | - | 3 - 4 | 534 | 71 | ||
| AK155117.1 | - | 3 - 4 | 636 | 71 | ||
| AK163601.1 | - | 3 - 4 | 626 | 71 | ||
| AK165014.1 | - | 3 - 4 | 635 | 71 | ||
| AK217153.1 | - | 1 - 2 | 86 | 71 | ||
| BC076638.1 | - | 3 - 4 | 626 | 30658833 | 71 | |
| DQ249870.1 | - | 3 - 4 | 372 | 71 | ||
| U54984.1 | - | 3 - 4 | 618 | 71 | ||
| X83536.2 | - | 3 - 4 | 544 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp14
Genomic Map