Assay Details
| Gene Symbol: Mef2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291191.1 | NP_001278120.1 | 2 - 3 | 206 | 95 | |
| NM_001291192.1 | NP_001278121.1 | 2 - 3 | 206 | 95 | ||
| GenBank mRNA | AK153751.1 | - | 2 - 3 | 207 | 95 | |
| BC019116.1 | - | 2 - 3 | 172 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001291191.1, NM_001291192.1 transcripts.
Set Membership: |
|

Mef2a
Genomic Map