Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400015758.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015758.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015758.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015760.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015760.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015760.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015762.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015762.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015762.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015761.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015761.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015761.mm.1 |
TC1400002075.mm.1 |
MTA TC |