Assay Details
| Gene Symbol: Myh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080728.2 | NP_542766.1 | 18 - 19 | 2099 | 82 | |
| GenBank mRNA | AK040973.1 | - | 18 - 19 | 2101 | 82 | |
| AK147031.1 | - | 18 - 19 | 2100 | 82 | ||
| AK157742.1 | - | 18 - 19 | 2100 | 82 | ||
| AK168677.1 | - | 2 - 3 | 130 | 82 | ||
| AK169198.1 | - | 18 - 19 | 2099 | 82 | ||
| AK169371.1 | - | 18 - 19 | 2099 | 82 | ||
| AY056464.1 | - | 18 - 19 | 2099 | 82 | ||
| BC121789.1 | - | 17 - 18 | 2055 | 40099448 | 82 | |
| BC158018.1 | - | 2176 | 9007324 | 82 | ||
| BC158070.1 | - | 18 - 19 | 2354 | 9007368 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myh7
Genomic Map