Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400015780.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015781.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015781.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015781.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015777.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015777.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015777.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015783.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015775.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015775.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015775.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015776.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015776.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015776.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015772.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015772.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015772.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015773.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015773.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015773.mm.1 |
TC1400002075.mm.1 |
MTA TC |