Assay Details
Gene Symbol: Pfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028376.3 | NP_082652.1 | 3 - 4 | 1217 | 70 | |
GenBank mRNA | AK013595.1 | - | 3 - 4 | 1219 | 70 | |
AY495949.1 | - | 4 - 5 | 760 | 70 | ||
BC116888.1 | - | 3 - 4 | 1061 | 40126207 | 70 | |
BC116890.1 | - | 4 - 5 | 648 | 40126209 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28202320 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139629 | NM_028376.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139629 | AK013595.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139629 | AY495949.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139629 | BC094614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139629 | BC116888.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139629 | NM_028376.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139629 | AK013595.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139629 | AY495949.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139629 | BC094614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139629 | BC116888.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746042 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637396 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | AK009291.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | AK122548.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | AK147464.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | AK147593.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | AK155296.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | AY178784.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | AY305656.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | AY305657.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | AY305658.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | BC094373.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | NM_001001602.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | NM_001114124.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731366 | NM_001114125.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366998 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853037 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643431 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29514 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140628 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139629 | NM_028376.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139629 | AK013595.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139629 | AY495949.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139629 | BC094614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139629 | BC116888.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139629 | NM_028376.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139629 | AK013595.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139629 | AY495949.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139629 | BC094614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139629 | BC116888.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139629 | NM_028376.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139629 | AK013595.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139629 | AY495949.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139629 | BC094614.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139629 | BC116888.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139629 | NM_028376.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139629 | AK013595.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139629 | AY495949.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139629 | BC094614.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139629 | BC116888.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202334 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202338 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202343 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202332 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202341 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202327 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202326 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202337 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202335 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202330 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202324 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28202321 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140628 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02450582 | - | - |
Whole Mouse Genome G4122A | A_52_P139629 | NM_028376.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139629 | AK013595.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139629 | AY495949.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139629 | BC094614.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139629 | BC116888.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139629 | NM_028376.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139629 | AK013595.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139629 | AY495949.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139629 | BC094614.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139629 | BC116888.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131932 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140628 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09906333 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09906332 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09906337 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28202320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28202360 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202330 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202351 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P10338965 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09906340 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202337 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202335 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202353 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202347 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202340 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202397 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09906333 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P10116915 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P10104769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202332 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28202319 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10394341 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394339 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394334 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394342 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394343 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394336 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394338 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394340 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394335 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394337 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394333 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428665_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10394331 | - | - |
Mouse Genome 430 2.0 Array | 1428665_at | - | - |
Mouse Genome 430 2.0 Array | 1428665_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200000438.mm.1 | NM_028376.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200000438.mm.1 | NM_028376.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200000438.mm.1 | TC1200000032.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200000437.mm.1 | NM_028376.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200000437.mm.1 | NM_028376.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200000437.mm.1 | TC1200000032.mm.1 | MTA TC |
Mouse Expression Set 430 | 1428665_at | - | - |
Mouse Expression Set 430 | 1428665_PM_at | - | - |
Murine Genome U74v2 Set | 166067_f_at | - | - |
Murine Genome U74v2 Set | 166067_PM_f_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428665_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428665_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 409394 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5860170 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2618400 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2618400 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2618398 | - | - |
