Assay Details
| Gene Symbol: Cdadc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168536.1 | NP_001162008.1 | 9 - 10 | 1599 | 70 | |
| GenBank mRNA | AK009280.1 | - | 9 - 10 | 1591 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdadc1
Genomic Map