Assay Details
| Gene Symbol: Zdhhc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029492.4 | NP_083768.4 | 8 - 9 | 824 | 102 | |
| GenBank mRNA | AK017158.1 | - | 8 - 9 | 825 | 102 | |
| AK031184.1 | - | 8 - 9 | 783 | 102 | ||
| AK049270.1 | - | 8 - 9 | 804 | 102 | ||
| AK140182.1 | - | 9 - 10 | 821 | 102 | ||
| AK151840.1 | - | 8 - 9 | 839 | 101 | ||
| AK163174.1 | - | 8 - 9 | 822 | 102 | ||
| AK170017.1 | - | 8 - 9 | 842 | 102 | ||
| AK170139.1 | - | 8 - 9 | 765 | 102 | ||
| AY668951.1 | - | 9 - 10 | 778 | 102 | ||
| BC019536.1 | - | 8 - 9 | 841 | 4039734 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc20
Genomic Map