Assay Details
| Gene Symbol: Pck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028994.2 | NP_083270.1 | 8 - 9 | 1595 | 118 | |
| GenBank mRNA | AK030501.1 | - | 8 - 9 | 1596 | 118 | |
| AK031612.1 | - | 8 - 9 | 1440 | 118 | ||
| AK034927.1 | - | 8 - 9 | 1474 | 118 | ||
| AK142561.1 | - | 2 - 3 | 347 | 118 | ||
| AK147972.1 | - | 8 - 9 | 1449 | 118 | ||
| AK165094.1 | - | 8 - 9 | 1473 | 118 | ||
| AK170242.1 | - | 8 - 9 | 1449 | 118 | ||
| AK218547.1 | - | 1 - 2 | 134 | 118 | ||
| BC010318.1 | - | 8 - 9 | 1458 | 3596227 | 118 | |
| BC011126.1 | - | 7 - 8 | 1757 | 118 | ||
| BC023437.1 | - | 6 - 7 | 1063 | 118 | ||
| BC028326.1 | - | 3 - 4 | 396 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pck2
Genomic Map