Assay Details
| Gene Symbol: Rabggta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019519.2 | NP_062392.1 | 5 - 6 | 967 | 86 | |
| GenBank mRNA | AF127656.1 | - | 6 - 7 | 815 | 86 | |
| AF127657.1 | - | 5 - 6 | 758 | 86 | ||
| AF127658.1 | - | 5 - 6 | 967 | 86 | ||
| AF127659.1 | - | 5 - 6 | 967 | 86 | ||
| AF127660.1 | - | 6 - 7 | 815 | 86 | ||
| AF127661.1 | - | 5 - 6 | 758 | 86 | ||
| AF127662.1 | - | 6 - 7 | 886 | 86 | ||
| AK002625.1 | - | 6 - 7 | 749 | 86 | ||
| AK146917.1 | - | 6 - 7 | 838 | 86 | ||
| BC012214.1 | - | 6 - 7 | 820 | 3967218 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabggta
Genomic Map