Assay Details
| Gene Symbol: Rabggta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019519.2 | NP_062392.1 | 8 - 9 | 1234 | 81 | |
| GenBank mRNA | AF127656.1 | - | 9 - 10 | 1082 | 81 | |
| AF127657.1 | - | 8 - 9 | 1025 | 81 | ||
| AF127658.1 | - | 8 - 9 | 1234 | 81 | ||
| AF127659.1 | - | 8 - 9 | 1234 | 81 | ||
| AF127660.1 | - | 9 - 10 | 1082 | 81 | ||
| AF127661.1 | - | 8 - 9 | 1025 | 81 | ||
| AF127662.1 | - | 9 - 10 | 1153 | 81 | ||
| AK002625.1 | - | 9 - 10 | 1016 | 81 | ||
| AK146917.1 | - | 9 - 10 | 1105 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabggta
Genomic Map