Assay Details
Gene Symbol: Tssk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253888.1 | NP_001240817.1 | 2 - 3 | 608 | 65 | |
NM_001253889.1 | NP_001240818.1 | 2 - 3 | 608 | 65 | ||
GenBank mRNA | AK006144.1 | - | 2 - 3 | 583 | 65 | |
DQ397205.1 | - | 2 - 3 | 508 | 65 | ||
DQ397206.1 | - | 2 - 3 | 508 | 65 | ||
EF127820.1 | - | 2 - 3 | 610 | 65 | ||
EF127821.1 | - | 2 - 3 | 609 | 65 | ||
EF127822.1 | - | 2 - 3 | 610 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
multicellular organism development
spermatogenesis
regulation of signal transduction
phosphorylation
cell differentiation
positive regulation of CREB transcription factor activity
intracellular signal transduction
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding
protein phosphorylation
multicellular organism development
spermatogenesis
regulation of signal transduction
phosphorylation
cell differentiation
positive regulation of CREB transcription factor activity
intracellular signal transduction
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101013 | EF127821.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101013 | EF127822.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101013 | EF127820.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101013 | AY461664.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101013 | DQ397204.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148149 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137825 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101013 | EF127821.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101013 | EF127822.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101013 | EF127820.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101013 | AY461664.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101013 | DQ397204.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619089 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619088 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619090 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619093 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619083 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619087 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619084 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137825 | - | - |
Whole Mouse Genome G4122A | A_51_P164788 | EF127821.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P164788 | EF127822.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P164788 | EF127820.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P164788 | AY461664.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P164788 | EF127819.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101013 | EF127821.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101013 | EF127822.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101013 | EF127820.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101013 | AY461664.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101013 | DQ397204.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132031 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137825 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11760146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619087 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619069 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760129 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619089 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11363679 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11169900 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619073 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760134 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619111 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619074 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619114 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619071 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619086 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11590579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11824072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619087 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619079 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619103 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619090 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619088 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619076 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619084 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10415357 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10415359 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10415355 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10415360 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10415362 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10415354 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10415361 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10415356 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438486_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443810_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432101_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10415353 | - | - |
Mouse Genome 430 2.0 Array | 1438486_at | - | - |
Mouse Genome 430 2.0 Array | 1443810_at | - | - |
Mouse Genome 430 2.0 Array | 1438486_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432101_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1443810_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432101_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71099_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71099_PM_at | - | - |
Mouse Expression Set 430 | 1438486_PM_at | - | - |
Mouse Expression Set 430 | 1438486_at | - | - |
Mouse Expression Set 430 | 1443810_PM_at | - | - |
Mouse Expression Set 430 | 1443810_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432101_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443810_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432101_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438486_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438486_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443810_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4060132 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004060132 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2630400 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2630400 | - | - |
mouse-6 v1.1 (Illumina) | scl46271.4.366_12-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4060132 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl46271.4.366_12-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060132 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2630400 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2630400 | - | - |
