Assay Details
| Gene Symbol: Dleu2 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028264.1 | 3 - 4 | 432 | 69 | |||
| GenBank mRNA | AF380423.1 | - | LIT3422 DLEU2 |
3 - 4 | 409 | 69 | |
| AF380425.1 | - | LIT3421 DLEU2 |
3 - 4 | 339 | 69 | ||
| AK042416.1 | - | 3 - 4 | 385 | 69 | |||
| AK080165.1 | - | 3 - 4 | 235 | 69 | |||
| AK084358.1 | - | 3 - 4 | 388 | 69 | |||
| AK134888.1 | - | 3 - 4 | 374 | 69 | |||
| AK165901.1 | - | 3 - 4 | 422 | 69 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 15a | Mir15a |
Mirn15a, mir-15a, mmu-mir-15a |
-327.0 | Sense | Chr. 14 - 61632027 - 61632110 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 27017 |
Gm27017 | 44.4kb | Anti-sense | Chr. 14 - 61680039 - 61686642 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120931 | AK165901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120931 | BB639368.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120931 | AK042416.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120931 | AK084358.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120931 | AF380423.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2200029 | AK033078.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2200034 | AK162803.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781104 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400721 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 897749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385984 | AK042167.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385984 | AK084270.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385984 | AK172484.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346471 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415594 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714813 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730721 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768677 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939649 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336022 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86813 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157997 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814593 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668434 | AK035475.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668434 | AK052776.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668434 | AK085411.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668434 | NM_028748.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206866 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682192 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215421 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918950 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109103 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863190 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223193 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225298 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293200 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680884 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721347 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815893 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375632 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8037 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50643 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168971 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97984 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99551 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88600 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176342 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163653 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178221 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2200029 | AK033078.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133229 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133153 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133174 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178221 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133130 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163653 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133192 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133182 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133200 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133241 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133142 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02863760 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133280 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176342 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08133211 | - | - |
| Whole Mouse Genome G4122A | A_51_P216015 | AK165901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216015 | AK134888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216015 | AK084358.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216015 | AF380423.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216015 | NR_028264.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489604 | AK033078.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P483974 | AK165901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P483974 | AK134888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P483974 | AK080165.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P483974 | AK084358.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P483974 | AF380423.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08133229 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163653 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178221 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176342 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08133142 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08133192 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02863760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11551557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11297588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11536700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11487629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11164950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11805700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11449623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11561868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11689105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11561861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11805701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11611297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11190416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11297587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29651214 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10420671 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427410_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456145_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427411_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10420670 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400016976.mm.1 | NR_028264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016976.mm.1 | NR_028264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016976.mm.1 | TC1400002195.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400016989.mm.1 | NR_028264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016989.mm.1 | NR_028264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016989.mm.1 | TC1400002195.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400016979.mm.1 | NR_028264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016979.mm.1 | NR_028264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016979.mm.1 | TC1400002195.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400016980.mm.1 | NR_028264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016980.mm.1 | NR_028264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016980.mm.1 | TC1400002195.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400016977.mm.1 | NR_028264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016977.mm.1 | NR_028264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016977.mm.1 | TC1400002195.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400016978.mm.1 | NR_028264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016978.mm.1 | NR_028264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016978.mm.1 | TC1400002195.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400016981.mm.1 | NR_028264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016981.mm.1 | NR_028264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016981.mm.1 | TC1400002195.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400016983.mm.1 | NR_028264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016983.mm.1 | NR_028264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016983.mm.1 | TC1400002195.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400016982.mm.1 | NR_028264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016982.mm.1 | NR_028264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016982.mm.1 | TC1400002195.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400017010.mm.1 | NR_028264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400017010.mm.1 | NR_028264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400017010.mm.1 | TC1400002195.mm.1 | MTA TC |

Dleu2
Genomic Map