Assay Details
| Gene Symbol: Prmt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013768.3 | NP_038796.2 | 5 - 6 | 71 | ||
| GenBank mRNA | AF167573.1 | - | 5 - 6 | 544 | 71 | |
| AK076346.1 | - | 5 - 6 | 71 | |||
| AK084106.1 | - | 4 - 5 | 71 | |||
| AK132414.1 | - | 5 - 6 | 71 | |||
| AK165165.1 | - | 5 - 6 | 71 | |||
| BC023905.1 | - | 5 - 6 | 623 | 5321370 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt5
Genomic Map