Assay Details
| Gene Symbol: Ripk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164107.1 | NP_001157579.1 | 1 - 2 | 245 | 69 | |
| GenBank mRNA | AK012177.1 | - | 1 - 2 | 245 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001164107.1 transcripts.
Set Membership: |
|

Ripk3
Genomic Map