Assay Details
| Gene Symbol: Ripk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164107.1 | NP_001157579.1 | 5 - 6 | 883 | 90 | |
| NM_001164108.1 | NP_001157580.1 | 5 - 6 | 981 | 90 | ||
| NM_019955.2 | NP_064339.2 | 5 - 6 | 900 | 90 | ||
| GenBank mRNA | AF178953.1 | - | 5 - 6 | 829 | 90 | |
| AK012177.1 | - | 5 - 6 | 881 | 90 | ||
| AK154505.1 | - | 5 - 6 | 893 | 90 | ||
| BC029210.1 | - | 5 - 6 | 851 | 3590770 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ripk3
Genomic Map