Assay Details
| Gene Symbol: Zfp36l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009549.2 | NP_001009549.1 | 1 - 2 | 2340 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2127959 | NM_001009549.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932107 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2127959 | NM_001009549.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32389028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32389002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32389000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32388987 | - | - |
| Whole Mouse Genome G4122A | A_52_P626357 | AY661338.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P626357 | NM_001009549.2 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14550655 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15055302 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14550654 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14550664 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14550666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32389003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32389002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32389000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32388961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32389034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15207270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14550664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14550670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32389003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32389028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14980401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32389000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15207273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14550668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15207261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32388964 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10604654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604655 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10604653 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00014270.mm.1 | NM_001009549.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00014270.mm.1 | NM_001009549.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00014270.mm.1 | TC0X00002246.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 802177 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250130 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2786954 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2786954 | - | - |

Zfp36l3
Genomic Map