Assay Details
| Gene Symbol: Vash2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144879.2 | NP_659128.2 | 6 - 7 | 1300 | 67 | |
| NR_027352.1 | 6 - 7 | 1167 | 67 | |||
| GenBank mRNA | AK045261.1 | - | 6 - 7 | 1096 | 67 | |
| AK078596.1 | - | 6 - 7 | 1169 | 67 | ||
| AK140989.1 | - | 6 - 7 | 1294 | 67 | ||
| AK163193.1 | - | 6 - 7 | 1076 | 67 | ||
| BC024141.1 | - | 6 - 7 | 1070 | 5149750 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vash2
Genomic Map