Assay Details
| Gene Symbol: Nek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010892.3 | NP_035022.2 | 5 - 6 | 940 | 85 | |
| GenBank mRNA | AF007247.1 | - | 5 - 6 | 854 | 85 | |
| AF013166.1 | - | 5 - 6 | 824 | 85 | ||
| AK077627.1 | - | 5 - 6 | 904 | 85 | ||
| AK147072.1 | - | 5 - 6 | 916 | 85 | ||
| AK164467.1 | - | 5 - 6 | 883 | 85 | ||
| AK169330.1 | - | 6 - 7 | 831 | 85 | ||
| AK170179.1 | - | 5 - 6 | 860 | 85 | ||
| AK171277.1 | - | 5 - 6 | 860 | 85 | ||
| AK171387.1 | - | 5 - 6 | 860 | 85 | ||
| BC010302.1 | - | 5 - 6 | 862 | 3597227 | 85 | |
| BC011316.1 | - | 5 - 6 | 860 | 3674001 | 85 | |
| BC057576.1 | - | 5 - 6 | 880 | 6810914 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek2
Genomic Map