Assay Details
Gene Symbol: Cnih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160211.1 | NP_001153683.1 | 2 - 3 | 949 | 71 | |
NM_001160212.1 | NP_001153684.1 | 2 - 3 | 949 | 71 | ||
NM_028408.3 | NP_082684.1 | 2 - 3 | 949 | 71 | ||
GenBank mRNA | AK013789.1 | - | 2 - 3 | 946 | 71 | |
AK033911.1 | - | 2 - 3 | 947 | 71 | ||
BC115640.1 | - | 2 - 3 | 575 | 40103001 | 71 | |
BC115641.1 | - | 2 - 3 | 575 | 40103002 | 71 | |
BG800984.1 | - | 2 - 3 | 379 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
