Assay Details
| Gene Symbol: Larp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138593.2 | NP_613059.2 | 11 - 12 | 1703 | 92 | |
| GenBank mRNA | BC029178.1 | - | 3 - 4 | 376 | 92 | |
| BC132266.1 | - | 3 - 4 | 374 | 92 | ||
| BC132268.1 | - | 3 - 4 | 374 | 92 | ||
| BM946683.1 | - | 5 - 6 | 571 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Larp7
Genomic Map