Assay Details
| Gene Symbol: Larp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138593.2 | NP_613059.2 | 2 - 3 | 358 | 92 | |
| GenBank mRNA | AK012772.1 | - | 2 - 3 | 348 | 92 | |
| AK015041.1 | - | 2 - 3 | 333 | 92 | ||
| AK082815.1 | - | 2 - 3 | 357 | 92 | ||
| AK162147.1 | - | 2 - 3 | 275 | 92 | ||
| BC020127.1 | - | 1 - 2 | 209 | 92 | ||
| BC023165.1 | - | 2 - 3 | 312 | 92 | ||
| BY269910.1 | - | 2 - 3 | 358 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Larp7
Genomic Map