Assay Details
| Gene Symbol: Larp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138593.2 | NP_613059.2 | 4 - 5 | 539 | 91 | |
| GenBank mRNA | AK012772.1 | - | 4 - 5 | 529 | 91 | |
| AK015041.1 | - | 4 - 5 | 514 | 91 | ||
| AK082815.1 | - | 4 - 5 | 538 | 91 | ||
| AK162147.1 | - | 4 - 5 | 456 | 91 | ||
| BC020127.1 | - | 3 - 4 | 390 | 91 | ||
| BC023165.1 | - | 4 - 5 | 493 | 91 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Larp7
Genomic Map