Assay Details
| Gene Symbol: Hmbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177338.5 | NP_796312.2 | 2 - 3 | 350 | 77 | |
| GenBank mRNA | AK052729.1 | - | 2 - 3 | 360 | 77 | |
| AK089782.1 | - | 2 - 3 | 351 | 77 | ||
| AK161592.1 | - | 2 - 3 | 350 | 77 | ||
| BC051457.1 | - | 2 - 3 | 353 | 6398463 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052729.1, AK089782.1, AK161592.1, BC051457.1, NM_177338.5 transcripts.
Set Membership: |
|

Hmbox1
Genomic Map