Assay Details
| Gene Symbol: Ddah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026993.3 | NP_081269.1 | 3 - 4 | 647 | 96 | |
| GenBank mRNA | AK010430.1 | - | 3 - 4 | 613 | 96 | |
| AK128909.1 | - | 3 - 4 | 202 | 96 | ||
| AK143507.1 | - | 3 - 4 | 664 | 96 | ||
| AK148907.1 | - | 3 - 4 | 648 | 96 | ||
| AK149185.1 | - | 3 - 4 | 299 | 96 | ||
| BC034505.1 | - | 3 - 4 | 598 | 5134831 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddah1
Genomic Map