Assay Details
| Gene Symbol: Ngly1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021504.3 | NP_067479.2 | 3 - 4 | 532 | 103 | |
| GenBank mRNA | AF250927.1 | - | 3 - 4 | 519 | 103 | |
| AK028248.1 | - | 3 - 4 | 533 | 103 | ||
| AK148931.1 | - | 3 - 4 | 535 | 103 | ||
| BC028961.1 | - | 3 - 4 | 486 | 5366023 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ngly1
Genomic Map