Assay Details
| Gene Symbol: Ngly1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021504.3 | NP_067479.2 | 6 - 7 | 1046 | 79 | |
| GenBank mRNA | AF250927.1 | - | 6 - 7 | 1033 | 79 | |
| AK028248.1 | - | 6 - 7 | 1047 | 79 | ||
| AK148931.1 | - | 6 - 7 | 1049 | 79 | ||
| AK169205.1 | - | 6 - 7 | 1034 | 79 | ||
| BC028961.1 | - | 6 - 7 | 1000 | 5366023 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ngly1
Genomic Map