Assay Details
| Gene Symbol: Oxsm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016905.1 | - | 2 - 3 | 286 | 151 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016905.1 transcripts.
Set Membership: |
|

Oxsm
Genomic Map