Assay Details
| Gene Symbol: 6530403H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032656.1 | - | 18 - 19 | 3111 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | polypyrimidine tract binding protein 2 | Ptbp2 |
Ptb2, brPTB, nPTB |
-670.5kb | Sense | Chr. 3 - 119718741 - 119783799 on NCBI Build GRCm38 |
| Downstream | RWD domain containing 3 | Rwdd3 |
2510027J23Rik, 3110037C01Rik, AI035684, AI662458, Rsume, X2CR1 |
696.9kb | Sense | Chr. 3 - 121155400 - 121171780 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

6530403H02Rik
Genomic Map