Assay Details
| Gene Symbol: 6530403H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032656.1 | - | 8 - 9 | 992 | 138 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | polypyrimidine tract binding protein 2 | Ptbp2 |
Ptb2, brPTB, nPTB |
-866.4kb | Sense | Chr. 3 - 119718741 - 119783799 on NCBI Build GRCm38 |
| Downstream | RWD domain containing 3 | Rwdd3 |
2510027J23Rik, 3110037C01Rik, AI035684, AI662458, Rsume, X2CR1 |
502.3kb | Sense | Chr. 3 - 121155400 - 121171780 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

6530403H02Rik
Genomic Map