Assay Details
| Gene Symbol: 9330178D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040553.1 | 1 - 2 | 87 | 62 | ||
| GenBank mRNA | AK034327.1 | - | 1 - 2 | 88 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine-rich repeats and IQ motif containing 3 | Lrriq3 |
4930438B07Rik, 4930511J15Rik, 4933403H06Rik, BB020367, Lrrc44 |
-782.1kb | Sense | Chr. 3 - 155093434 - 155194278 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930570G19 gene | 4930570G19Rik | C630015G22 | 559.3kb | Anti-sense | Chr. 3 - 156546753 - 156561746 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

9330178D15Rik
Genomic Map