Assay Details
| Gene Symbol: Gm26544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033312.1 | - | 2 - 3 | 360 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | vascular cell adhesion molecule 1 | Vcam1 |
CD106, Vcam-1 |
-30.6kb | Anti-sense | Chr. 3 - 116110020 - 116129688 on NCBI Build GRCm38 |
| Downstream | G-protein coupled receptor 88 | Gpr88 |
AW061286, Strg |
88.9kb | Anti-sense | Chr. 3 - 116249654 - 116253498 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm26544
Genomic Map