Assay Details
| Gene Symbol: Gm26544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033312.1 | - | 4 - 5 | 616 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | vascular cell adhesion molecule 1 | Vcam1 |
CD106, Vcam-1 |
-33.0kb | Anti-sense | Chr. 3 - 116110020 - 116129688 on NCBI Build GRCm38 |
| Downstream | G-protein coupled receptor 88 | Gpr88 |
AW061286, Strg |
86.3kb | Anti-sense | Chr. 3 - 116249654 - 116253498 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26544
Genomic Map