Assay Details
| Gene Symbol: Kif26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161665.1 | NP_001155137.1 | 2 - 3 | 908 | 86 | |
| GenBank mRNA | AB355846.1 | - | 2 - 3 | 908 | 86 | |
| AK052046.1 | - | 2 - 3 | 909 | 86 | ||
| BC120791.1 | - | 2 - 3 | 571 | 40129714 | 86 | |
| BC137689.1 | - | 2 - 3 | 571 | 8860708 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif26b
Genomic Map