Assay Details
| Gene Symbol: 4930438E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045961.1 | 4 - 5 | 250 | 76 | ||
| GenBank mRNA | AK015336.1 | - | 4 - 5 | 251 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6878 | Gm6878 | EG628416 | -37.5kb | Sense | Chr. 14 - 67304888 - 67314711 on NCBI Build GRCm38 |
| Downstream | predicted gene 10860 | Gm10860 | ENSMUSG00000075548 | 73.4kb | Sense | Chr. 14 - 67430194 - 67437171 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930438E09Rik
Genomic Map