Assay Details
| Gene Symbol: Gm17116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041104.1 | - | 2 - 3 | 161 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protease, serine 52 |
Prss52 |
1700049K14Rik, Tesp3 |
-81.0 | Anti-sense | Chr. 14 - 64104323 - 64113751 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9630015K15 gene | 9630015K15Rik | 1.8kb | Anti-sense | Chr. 14 - 64116233 - 64127625 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm17116
Genomic Map